1-(2-Phthalimidobutyryl)chloride ≥95%

Supplier: Ambeed
Danger

A348616-1G A348616-5G
77901-569EA 86 USD
77901-569 77901-329
1-(2-Phthalimidobutyryl)chloride ≥95%
1-(2-Phthalimidobutyryl)chloride

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO₃
MDL Number: MFCD02656536
CAS Number: 10314-06-4
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR