rel-(3aR,5r,6aS)-tert-butyl 5-aminohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
cis-5-amino-2-boc-hexahydro-cyclopenta[c]pyrrole
A348596-100MG
A348596-250MG
77685-988EA
75
USD
77685-988
77685-989
rel-(3aR,5r,6aS)-tert-butyl 5-aminohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate ≥98%
rel-(3aR,5r,6aS)-tert-butyl 5-aminohexahydrocyclopenta[c]pyrrole-2(1H)-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₂₂N₂O₂ MW: 226.32 g/mol |
MDL Number:
MFCD29056567 CAS Number: 863600-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...