(3R,4R,5S)-Ethyl 4-acetamido-5-amino-3-(pentan-3-yloxy)cyclohex-1-enecarboxylate ≥97%

Supplier: Ambeed
Warning

Synonyms: Oseltamivir

A348556-100MG A348556-1G A348556-250MG
77601-556EA 48.18 USD
77601-556 77601-558 77668-596
(3R,4R,5S)-Ethyl 4-acetamido-5-amino-3-(pentan-3-yloxy)cyclohex-1-enecarboxylate ≥97%
(3R,4R,5S)-Ethyl 4-acetamido-5-amino-3-(pentan-3-yloxy)cyclohex-1-enecarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₈N₂O₄
MW: 312.40 g/mol
MDL Number: MFCD00953939
CAS Number: 196618-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR