3-Bromo-1-(triisopropylsilyl)indole ≥95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-1-(triisopropylsilyl)-1H-indole

A354987-5G A354987-1G
77572-044EA 132.87 USD
77572-044 77616-556
3-Bromo-1-(triisopropylsilyl)indole ≥95%
3-Bromo-1-(triisopropylsilyl)indole

Precaución: for Research Use Only

Formula: C₁₇H₂₆BrNSi
MW: 352.39 g/mol
MDL Number: MFCD02093441
CAS Number: 148249-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR