5-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥96%

Supplier: Ambeed
Warning

A354850-1G A354850-5G
77892-726EA 171 USD
77892-726 77892-727
5-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥96%
5-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂O₂
MDL Number: MFCD08059289
CAS Number: 1187932-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR