4-Amino-6-chloropyridazin-3(2H)-one ≥98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A347620-100MG
A347620-250MG
A347620-1G
77684-025EA
22
USD
77684-025
77684-026
77892-716
4-Amino-6-chloropyridazin-3(2H)-one ≥98%, Ambeed.Inc
4-Amino-6-chloropyridazin-3(2H)-one
Precaución: For research use only.
Formula:
C₄H₄ClN₃O |
MDL Number:
MFCD11109440 CAS Number: 14704-64-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...