Ethyl 3-(4-formylphenyl)propanoate ≥97%

Supplier: Ambeed
Warning

A347483-250MG A347483-100MG
77668-594EA 305 USD
77668-594 77668-592
Ethyl 3-(4-formylphenyl)propanoate ≥97%
Ethyl 3-(4-formylphenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.23 g/mol
MDL Number: MFCD08234825
CAS Number: 445483-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR