4-Chloro-3-iodotoluene ≥95%

Supplier: Ambeed
Warning

A347420-5G A347420-1G
77901-077EA 91 USD
77901-077 77901-879
4-Chloro-3-iodotoluene ≥95%
4-Chloro-3-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClI
MDL Number: MFCD09753727
CAS Number: 2401-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR