5-Nitro-2-furaldehyde diacetate ≥97%

Supplier: Ambeed
Warning

A354476-500G A354476-25G
77572-042EA 41 USD
77572-042 77885-934
5-Nitro-2-furaldehyde diacetate ≥97%
5-Nitro-2-furaldehyde diacetate

Precaución: for Research Use Only

Formula: C₉H₉NO₇
MDL Number: MFCD00003244
CAS Number: 92-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR