3-Bromo-4'-fluorobiphenyl ≥98%

Supplier: Ambeed
Warning

A354164-1G A354164-5G
77900-802EA 24 USD
77900-802 77900-602
3-Bromo-4'-fluorobiphenyl ≥98%
3-Bromo-4'-fluorobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrF
MDL Number: MFCD04038228
CAS Number: 10540-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR