4-(2-Methyl-1-(4-methylpiperazin-1-yl)-1-oxopropan-2-yl)-N-(6-(pyridin-4-yl)imidazo[1,2-a]pyridin-2-yl)benzamide 98+%

Supplier: Ambeed
Warning

Synonyms: CLK-IN-T3

A353812-50MG A353812-250MG A353812-100MG
77748-701EA 389 USD
77748-701 77748-700 77748-699
4-(2-Methyl-1-(4-methylpiperazin-1-yl)-1-oxopropan-2-yl)-N-(6-(pyridin-4-yl)imidazo[1,2-a]pyridin-2-yl)benzamide 98+%
4-(2-Methyl-1-(4-methylpiperazin-1-yl)-1-oxopropan-2-yl)-N-(6-(pyridin-4-yl)imidazo[1,2-a]pyridin-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₂₈H₃₀N₆O₂
MW: 482.58 g/mol
MDL Number: MFCD31689325
CAS Number: 2109805-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR