4-Chloro-3,5-dimethylphenylboronic acid pinacol ester ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethyl-4-chlorophenylboronic acid pinacol ester, 2-(4-Chloro-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A353468-1G A353468-100MG A353468-250MG A353468-5G
77684-042EA 152 USD
77684-042 77885-931 77885-932 77885-933
4-Chloro-3,5-dimethylphenylboronic acid pinacol ester ≥98%, Ambeed.Inc
4-Chloro-3,5-dimethylphenylboronic acid pinacol ester

Precaución: For research use only.

Formula: C₁₄H₂₀BClO₂
MDL Number: MFCD12405360
CAS Number: 1111096-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR