Coomassie® brilliant blue R-250 ≥98%

Supplier: Ambeed
Danger

Synonyms: Serva Blue R, Brillant blue R, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulphophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulpho-benzenemethanaminium inner salt, sodium salt (1:1), Coomassie Blue R 250, Brilliant Cyanine 6B, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt, Acid Blue 83, Brilliant Blue R250, Brilliant Blue R, Brilliant indocyanine 6B

A352910-5G A352910-25G
77601-584EA 38.95 USD
77601-584 77626-682
Coomassie® brilliant blue R-250 ≥98%
Coomassie® brilliant blue R-250

Precaución: for Research Use Only

Formula: C₄₅H₄₄N₃NaO₇S₂
MW: 825.98 g/mol
MDL Number: MFCD00041762
CAS Number: 6104-59-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR