Boc-Cys(Mbzl)-OH ≥98%

Supplier: Ambeed
Warning

Synonyms: N-α-t.-Boc-S-p-methylbenzyl-L-cysteine, Boc-Cys(pMeBzl)-OH

A352819-25G A352819-5G A352819-100G A352819-1G
77668-634EA 47 USD
77668-634 77668-636 77668-630 77668-632
Boc-Cys(Mbzl)-OH ≥98%
Boc-Cys(Mbzl)-OH

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₄S
MW: 325.423 g/mol
MDL Number: MFCD00038517
CAS Number: 61925-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR