Fmoc-3-bromo-L-phenylalanine ≥97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(3-Br)-OH

A352779-250MG A352779-25G
77872-526EA 27 USD
77872-526 77872-527
Fmoc-3-bromo-L-phenylalanine ≥97%
Fmoc-3-bromo-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀BrNO₄
MW: 466.34 g/mol
MDL Number: MFCD01311744
CAS Number: 220497-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR