(1-(tert-Butoxycarbonyl)-4-methoxy-1H-indol-2-yl)boronic acid 98+%

Supplier: Ambeed
Warning

A345592-1G A345592-250MG A345592-5G
77885-907EA 60 USD
77885-907 77885-908 77885-909
(1-(tert-Butoxycarbonyl)-4-methoxy-1H-indol-2-yl)boronic acid 98+%
(1-(tert-Butoxycarbonyl)-4-methoxy-1H-indol-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₅
MW: 291.11 g/mol
MDL Number: MFCD11616286
CAS Number: 1000068-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR