Cesium bicarbonate ≥99%

Supplier: Ambeed
Warning

A344129-500G A344129-5G
77626-658EA 1398.74 USD
77626-658 77879-739
Cesium bicarbonate ≥99%
Cesium bicarbonate

Precaución: for Research Use Only

Formula: CHCsO₃
MW: 193.92 g/mol
MDL Number: MFCD00010961
CAS Number: 29703-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR