1-(Isoquinolin-3-yl)ethanone ≥96%

Supplier: Ambeed
Warning

A343820-250MG A343820-100MG A343820-1G
77685-983EA 58 USD
77685-983 77892-702 77892-703
1-(Isoquinolin-3-yl)ethanone ≥96%
1-(Isoquinolin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₉NO
MDL Number: MFCD06658291
CAS Number: 91544-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR