Potassium tetrakis(4-chlorophenyl)borate ≥98%
Supplier: Ambeed
|
Warning
|
A343689-1G
A343689-250MG
77668-570EA
107
USD
77668-570
77668-572
Potassium tetrakis(4-chlorophenyl)borate ≥98%
Potassium tetrakis(4-chlorophenyl)borate
Precaución: for Research Use Only
Formula:
C₂₄H₁₆BCl₄K |
MDL Number:
MFCD00043105 CAS Number: 14680-77-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...