(S)-5-Methylpyrrolidin-2-one ≥98%

Supplier: Ambeed
Warning

A344821-100MG A344821-250MG
77748-675EA 35 USD
77748-675 77748-676
(S)-5-Methylpyrrolidin-2-one ≥98%
(S)-5-Methylpyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₅H₉NO
MW: 99.13 g/mol
MDL Number: MFCD19216982
CAS Number: 1558-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR