(R)-(-)-Tetrahydrofurfuryl alcohol ≥98%

Supplier: Ambeed
Warning

Synonyms: (R)-Tetrahydro-2-furanmethanol, (R)-(Tetrahydrofuran-2-yl)methanol

A344649-1G A344649-250MG
77572-002EA 46.35 USD
77572-002 77612-970
(R)-(-)-Tetrahydrofurfuryl alcohol ≥98%
(R)-(-)-Tetrahydrofurfuryl alcohol

Precaución: for Research Use Only

Formula: C₅H₁₀O₂
MW: 102.133 g/mol
MDL Number: MFCD03093085
CAS Number: 22415-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR