2,2-Difluorocyclopentan-1-amine hydrochloride ≥95%

Supplier: Ambeed
Warning

A342869-100MG A342869-1G A342869-250MG
77900-651EA 46 USD
77900-651 77901-523 77900-784
2,2-Difluorocyclopentan-1-amine hydrochloride ≥95%
2,2-Difluorocyclopentan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClF₂N
MDL Number: MFCD18909233
CAS Number: 921599-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR