4-Amino-2-(methylthio)pyrimidine-5-carboxylic acid ≥98%
Supplier: Ambeed
|
Warning
|
A334798-1G
A334798-5G
77684-000EA
22
USD
77684-000
77684-001
4-Amino-2-(methylthio)pyrimidine-5-carboxylic acid ≥98%
4-Amino-2-(methylthio)pyrimidine-5-carboxylic acid
Precaución: For research use only.
Formula:
C₆H₇N₃O₂S MW: 185.2 g/mol |
MDL Number:
MFCD00186102 CAS Number: 771-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...