3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: 3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid, 3-{[(tert-butoxy)carbonyl]amino}cyclobutane-1-carboxylic acid

A334694-1G A334694-5G A334694-250MG A334694-25G A334694-10G
77612-918EA 23.69 USD
77612-918 77612-920 77672-708 77672-710 77672-706
3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid ≥97%
3-(tert-Butoxycarbonylamino)cyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
MDL Number: MFCD09864181
CAS Number: 946152-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR