Ethyl 2-(4-hydroxyphenyl)-2-oxoacetate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A342755-1G A342755-250MG
77684-016EA 91 USD
77684-016 77892-700
Ethyl 2-(4-hydroxyphenyl)-2-oxoacetate ≥95%, Ambeed.Inc
Ethyl 2-(4-hydroxyphenyl)-2-oxoacetate

Precaución: For research use only.

Formula: C₁₀H₁₀O₄
MDL Number: MFCD20260326
CAS Number: 70080-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR