Micafungin sodium ≥98%

Supplier: Ambeed
Warning

Synonyms: Sodium 5-((1S,2S)-2-((2R,6S,9S,11R,12R,14aS,15S,16S,20S,23S,25aS)-20-((R)-3-amino-1-hydroxy-3-oxopropyl)-2,8,11,12,14,15,22,25-octahydroxy-6-((R)-1-hydroxyethyl)-16-methyl-5,19-dioxo-9-(4-(5-(4-(pentyloxy)phenyl)isoxazol-3-yl)benzamido)-2,3,5,6,9,10,11,12,14a,15,16,17,19,20,23,25a-hexadecahydro-1H-dipyrrolo[2,1-c:2',1'-l][1,4,7,10,13,16]hexaazacyclohenicosin-23-yl)-1,2-dihydroxyethyl)-2-hydroxyphenyl sulphate

A334667-10MG A334667-1MG A334667-250MG A334667-100MG
77616-470EA 31.93 USD
77616-470 77616-472 77746-652 77746-651
Micafungin sodium ≥98%
Micafungin sodium

Precaución: for Research Use Only

Formula: C₅₆H₇₀N₉NaO₂₃S
MW: 1292.256 g/mol
MDL Number: MFCD08067752
CAS Number: 208538-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR