Ethyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥98%

Supplier: Ambeed
Warning

A334400-1G A334400-250MG A334400-5G
77883-918EA 189 USD
77883-918 77883-919 77883-920
Ethyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥98%
Ethyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₆H₂₃BO₄
MDL Number: MFCD12026076
CAS Number: 1150271-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR