Benzo[d]oxazole-6-carbaldehyde ≥97%

Supplier: Ambeed
Warning

A341974-250MG A341974-100MG
77593-998EA 120.95 USD
77593-998 77612-958
Benzo[d]oxazole-6-carbaldehyde ≥97%
Benzo[d]oxazole-6-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅NO₂
MW: 147.13 g/mol
MDL Number: MFCD11040758
CAS Number: 865449-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR