1H-Pyrrolo[2,3-c]pyridine-3-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A333691-1G A333691-100MG A333691-5G
77593-968EA 86.1 USD
77593-968 77883-916 77883-917
1H-Pyrrolo[2,3-c]pyridine-3-carboxylic acid ≥97%
1H-Pyrrolo[2,3-c]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD11845479
CAS Number: 67058-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR