3-Azabicyclo[3.1.1]heptane hydrochloride ≥98%

Supplier: Ambeed
Warning

A341729-250MG A341729-50MG
77571-992EA 400.67 USD
77571-992 77684-015
3-Azabicyclo[3.1.1]heptane hydrochloride ≥98%
3-Azabicyclo[3.1.1]heptane hydrochloride

Precaución: For research use only.

Formula: C₆H₁₂ClN
MW: 133.62 g/mol
MDL Number: MFCD23144086
CAS Number: 1427380-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR