1-Boc-3-(methylaminomethyl)azetidine ≥98%

Supplier: Ambeed
Danger

A333404-1G A333404-100MG A333404-250MG A333404-5G
77601-498EA 257.28 USD
77601-498 77668-514 77668-516 77872-483
1-Boc-3-(methylaminomethyl)azetidine ≥98%
1-Boc-3-(methylaminomethyl)azetidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD08061965
CAS Number: 1049730-81-5
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR