Fmoc-His(Bzl)-OH ≥95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1-benzyl-1H-imidazol-4-yl)propanoic acid

A333217-1G A333217-250MG A333217-5G
77879-709EA 56 USD
77879-709 77879-710 77879-711
Fmoc-His(Bzl)-OH ≥95%
Fmoc-His(Bzl)-OH

Precaución: for Research Use Only

Formula: C₂₈H₂₅N₃O₄
MW: 467.52 g/mol
MDL Number: MFCD00038537
CAS Number: 84030-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR