Benzyl-2-acetamido-4,6-O-benzylidene-2-deoxy-α-D-glucopyranoside ≥95%
Supplier: Ambeed
|
Warning
|
A340930-25G
A340930-5G
77878-043EA
338
USD
77878-043
77878-044
Benzyl-2-acetamido-4,6-O-benzylidene-2-deoxy-α-D-glucopyranoside ≥95%
Benzyl-2-acetamido-4,6-O-benzylidene-2-deoxy-alpha-D-glucopyranoside
Precaución: for Research Use Only
Formula:
C₂₂H₂₅NO₆ |
MDL Number:
MFCD00143679 CAS Number: 13343-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...