Fmoc-Orn(ivDde)-OH ≥97%

Supplier: Ambeed
Warning

A340912-5G A340912-1G
77571-988EA 436.72 USD
77571-988 77684-013
Fmoc-Orn(ivDde)-OH ≥97%
Fmoc-Orn(ivDde)-OH

Precaución: For research use only.

Formula: C₃₃H₄₀N₂O₆
MW: 560.68 g/mol
MDL Number: MFCD03788161
CAS Number: 1198321-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR