4-Hydroxy-3-nitrobenzoic acid ≥97%

Supplier: Ambeed
Warning

A333149-25G A333149-5G
77571-960EA 22 USD
77571-960 77883-914
4-Hydroxy-3-nitrobenzoic acid ≥97%
4-Hydroxy-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₅
MDL Number: MFCD00007119
CAS Number: 616-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR