6-Chloro-5-methoxypyridine-3-boronic acid ≥98%

Supplier: Ambeed
Warning

A340683-100MG A340683-1G A340683-250MG
77901-209EA 46 USD
77901-209 77900-985 77900-546
6-Chloro-5-methoxypyridine-3-boronic acid ≥98%
6-Chloro-5-methoxypyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₃
MDL Number: MFCD13189015
CAS Number: 1548827-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR