4-(Dimethylsulfamoyl)benzene-1-sulfonyl chloride ≥95%

Supplier: Ambeed
Danger

A340511-1G A340511-250MG A340511-5G
77900-617EA 434 USD
77900-617 77901-694 77900-619
4-(Dimethylsulfamoyl)benzene-1-sulfonyl chloride ≥95%
4-(Dimethylsulfamoyl)benzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₄S₂
MDL Number: MFCD06340105
CAS Number: 677782-39-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR