2,6-Diaminopimelic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: 1,5-Diaminoheptanedioic Acid, 2,6-Diaminoheptanedioic acid

A332870-100MG A332870-5G
77685-950EA 59 USD
77685-950 77685-951
2,6-Diaminopimelic acid ≥97%
2,6-Diaminopimelic acid

Precaución: for Research Use Only

Formula: C₇H₁₄N₂O₄
MW: 190.20 g/mol
MDL Number: MFCD00002637
CAS Number: 583-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR