tert-Butyl ((3S,4R)-4-aminotetrahydrofuran-3-yl)carbamate ≥97%

Supplier: Ambeed
Warning

A340225-100MG A340225-1G A340225-250MG
77593-986EA 128.13 USD
77593-986 77593-988 77593-990
tert-Butyl ((3S,4R)-4-aminotetrahydrofuran-3-yl)carbamate ≥97%
tert-Butyl ((3S,4R)-4-aminotetrahydrofuran-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₃
MW: 202.2508 g/mol
MDL Number: MFCD27956985
CAS Number: 1932043-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR