3,5-Dibromopyridazine ≥96%

Supplier: Ambeed
Warning

A339974-1G A339974-250MG
77872-500EA 329 USD
77872-500 77872-501
3,5-Dibromopyridazine ≥96%
3,5-Dibromopyridazine

Precaución: for Research Use Only

Formula: C₄H₂Br₂N₂
MDL Number: MFCD13190294
CAS Number: 1196155-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR