4,4'-Dihydroxy-2,2'-bipyridine ≥97%

Supplier: Ambeed
Warning

A339806-250MG A339806-100MG A339806-1G A339806-5G
77571-984EA 55 USD
77571-984 77612-944 77612-946 77612-948
4,4'-Dihydroxy-2,2'-bipyridine ≥97%
4,4'-Dihydroxy-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD08704189
CAS Number: 90770-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR