(R)-Methyl 2-hydroxy-3-methylbutanoate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A339611-25G A339611-5G
77684-007EA 1321 USD
77684-007 77890-678
(R)-Methyl 2-hydroxy-3-methylbutanoate ≥95%, Ambeed.Inc
(R)-Methyl 2-hydroxy-3-methylbutanoate

Precaución: For research use only.

Formula: C₆H₁₂O₃
MDL Number: MFCD12964323
CAS Number: 90244-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR