6-Fluoro-DL-tryptophan ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Amino-3-(6-fluoro-1H-indol-3-yl)propanoic acid, 6-Fluoro-DL-tryptophan
A339425-1G
A339425-250MG
A339425-5G
A339425-100MG
77668-558EA
159
USD
77668-558
77668-560
77668-562
77879-729
6-Fluoro-DL-tryptophan ≥95%
6-Fluoro-DL-tryptophan
Precaución: for Research Use Only
Formula:
C₁₁H₁₁FN₂O₂ |
MDL Number:
MFCD00005653 CAS Number: 7730-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...