1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R)-quinolin-4-yl(5-vinylquinuclidin-2-yl)methyl)thiourea ≥97% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with TBC

A338678-1G A338678-250MG
77593-978EA 161.95 USD
77593-978 77684-006
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R)-quinolin-4-yl(5-vinylquinuclidin-2-yl)methyl)thiourea ≥97% stabilized
1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R)-quinolin-4-yl(5-vinylquinuclidin-2-yl)methyl)thiourea

Precaución: For research use only.

Formula: C₂₈H₂₆F₆N₄S
MW: 564.59 g/mol
MDL Number: MFCD23704929
CAS Number: 851477-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR