(E)-(2R,3S,4R,5R,6R)-6-(3,4-Dihydroxyphenethoxy)-4,5-dihydroxy-2-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3-yl 3-(3,4-dihydroxyphenyl)acrylate 98+%

Supplier: Ambeed
Warning

Synonyms: Forsythoside A

A338187-100MG A338187-250MG A338187-50MG A338187-1G
77668-542EA 132 USD
77668-542 77668-544 77668-546 77879-726
(E)-(2R,3S,4R,5R,6R)-6-(3,4-Dihydroxyphenethoxy)-4,5-dihydroxy-2-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3-yl 3-(3,4-dihydroxyphenyl)acrylate 98+%
(E)-(2R,3S,4R,5R,6R)-6-(3,4-Dihydroxyphenethoxy)-4,5-dihydroxy-2-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3-yl 3-(3,4-dihydroxyphenyl)acrylate

Precaución: for Research Use Only

Formula: C₂₉H₃₆O₁₅
MDL Number: MFCD08460220
CAS Number: 79916-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR