(S)-2,2'-Dimethoxy-1,1'-binaphthyl ≥98%, ee 99%

Supplier: Ambeed
Warning

A330851-25G A330851-5G
77883-905EA 333 USD
77883-905 77883-906
(S)-2,2'-Dimethoxy-1,1'-binaphthyl ≥98%, ee 99%
(S)-2,2'-Dimethoxy-1,1'-binaphthyl

Precaución: for Research Use Only

Formula: C₂₂H₁₈O₂
MDL Number: MFCD00091146
CAS Number: 75640-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR