4-[(9H-Fluoren-9-yloxy)carbonyl]-1-(tert-butoxycarbonyl)piperazyl-2-acetic acid ≥98%
Supplier: Ambeed
|
Warning
|
A337979-1G
A337979-5G
77881-932EA
70
USD
77881-932
77881-933
4-[(9H-Fluoren-9-yloxy)carbonyl]-1-(tert-butoxycarbonyl)piperazyl-2-acetic acid ≥98%
4-[(9H-Fluoren-9-yloxy)carbonyl]-1-(tert-butoxycarbonyl)piperazyl-2-acetic acid
Precaución: for Research Use Only
Formula:
C₂₆H₃₀N₂O₆ |
MDL Number:
MFCD00674206 CAS Number: 183742-34-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...