5,6-Dichloropyridine-3-sulfonyl chloride ≥98%

Supplier: Ambeed
Danger

A330846-1G A330846-250MG
77885-867EA 389 USD
77885-867 77885-868
5,6-Dichloropyridine-3-sulfonyl chloride ≥98%
5,6-Dichloropyridine-3-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₅H₂Cl₃NO₂S
MDL Number: MFCD03541048
CAS Number: 98121-40-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR