3-Bromo-4-(difluoromethoxy)pyridine ≥96%

Supplier: Ambeed
Warning

A330522-250MG A330522-1G
77748-648EA 174 USD
77748-648 77748-647
3-Bromo-4-(difluoromethoxy)pyridine ≥96%
3-Bromo-4-(difluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrF₂NO
MW: 224 g/mol
MDL Number: MFCD13185845
CAS Number: 1214377-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR