2-(7-Hydroxy-5-phenethyl-[1,2,4]triazolo[1,5-a]pyridine-8-carboxamido)acetic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: Enarodustat

A337507-50MG A337507-100MG
77748-667EA 250 USD
77748-667 77748-666
2-(7-Hydroxy-5-phenethyl-[1,2,4]triazolo[1,5-a]pyridine-8-carboxamido)acetic acid ≥97%
2-(7-Hydroxy-5-phenethyl-[1,2,4]triazolo[1,5-a]pyridine-8-carboxamido)acetic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₄O₄
MW: 340.33 g/mol
MDL Number: MFCD31692356
CAS Number: 1262132-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR